Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Nitrate, 0.045 N, Ricca Chemical
SDP

Catalog No. R696500010F
Click to view available options
Quantity:
4 L
10 L
Packaging:
Black Poly Bottle
Cubitainer

Chemical Identifiers

CAS 7761-88-8, 7732-18-5
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Color Colorless
CAS Min % 97.0, 0.74
CAS Max % 100.0, 0.77
Grade Laboratory
Packaging Cubitainer
pH ∼6
Quantity 10 L
Physical Form Liquid
Concentration or Composition Notes 0.045Normal
Chemical Name or Material Silver Nitrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.