Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Silver Nitrate, 0.100 N (N/10) in Denatured Alcohol, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Poly Bottle
Black Poly Bottle
Specifications
Specifications
| Color | Colorless to Tan |
| CAS Min % | 2.04, 96.0 |
| CAS Max % | 2.12, 100.0 |
| Grade | Laboratory |
| Packaging | Amber Poly Bottle |
| Solvent or Matrix | Denatured Alcohol |
| Quantity | 1 L |
| Physical Form | Liquid |
| Concentration or Composition Notes | 0.100Normal |
| Chemical Name or Material | Silver Nitrate |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction