Learn More
Silver nitrate, 0.1N Standardized Solution
Description
Silver nitrate, 1.0N Standardized Solution is used for volumetric analysis in laboratories. It is widely used as an analytical reagent owing to its immediate reaction with halide ions forming insoluble precipitate of silver(I) halides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless |
| Physical Form | Liquid |
| Quantity | 4 L |
| Solubility Information | Miscible with water,alcohol and aqueous ammonia. |
| Formula Weight | 169.87 |
| Concentration or Composition (by Analyte or Components) | Silver nitrate: 1.7%; Water: 98.3% |
| Odor | Odorless |
| Sensitivity | Light sensitive |
| Appearance | Colourless liquid |
| Vapor Pressure | 23 hPa (17mm Hg) at 20°C |
| Show More |
Safety and Handling
| Hazard Category | H315-H319 |
| Precautionary Statement | P234-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362 |
| DOTInformation | DOT Class: 8; Packaging Group: III |
| EINECSNumber | 231-853-9 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.