Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Nitrate, 0.500 Normal (N/2), Reagents

Catalog No. 01334017
Change view
Click to view available options
Quantity:
1 L
4 L
Catalog No. Quantity
01-334-017 1 L
01-334-016 4 L
2 options

Catalog No. 01-334-017

Supplier: Reagents Holdings Llc CS0047001C

Only null left
Add to Cart
Add to Cart

Silver Nitrate, 0.500 Normal (N/2), Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 7761-88-8
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Nitric Acid silver salt
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Colorless
Physical Form Liquid
Quantity 1 L
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.0704
Chemical Name or Material Silver Nitrate, 0.500 Normal (N/2)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.