Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Nitrate, 0.6 N, Unstandardized, Reagents

Catalog No. 01333924
Change view
Click to view available options
Quantity:
10 L
15 gal.
Catalog No. Quantity
01-333-924 10 L
01-333-923 15 gal.
2 options

Catalog No. 01-333-924

Supplier: Reagents Holdings Llc CS02090010F

Only null left
Add to Cart
Add to Cart

Silver Nitrate, 0.6 N, Unstandardized, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Silver Nitrate, 0.6 N, Unstandardized
Boiling Point 100°C
CAS 7732-18-5, 7761-88-8
Color Colorless
Physical Form Liquid
Quantity 10 L
Molecular Formula AgNO3/H2O
Synonym Nitric Acid silver salt
Grade Reagent
Packaging Cubitainer
Specific Gravity 1.079
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.