Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Nitrate, 2% (w/v) in 2% (v/v) Nitric Acid, Ricca Chemical
SDP

Catalog No. R6826200500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R6826200500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R6826200500 Supplier Ricca Chemical Company Supplier No. R6826200500B
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7761-88-8 , 7732-18-5 , 7697-37-2
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Color Colorless
CAS Min % 1.87, 93.0, 2.8
CAS Max % 1.94, 97.0, 2.92
Percent Purity 2%
Grade Laboratory
Packaging Amber Poly Bottle
Solvent or Matrix 2% (v/v) Nitric Acid
pH <1
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 2% (w/v)
Chemical Name or Material Silver Nitrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.