Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Silver nitrate, 99.85%, for analysis
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis

Specifications
Specifications
Boiling Point | 444.0°C |
Melting Point | 212.0°C |
Color | White |
Physical Form | Crystalline Powder and/or Crystals |
Quantity | 2.5 kg |
Assay Percent Range | 99.85% |
Linear Formula | AgNO3 |
Fieser | 01,1008; 02,366; 03,252; 04,429; 05,582; 07,321; 09,411; 10,350; 11,268; 12,257; 14,350; 15,39 |
Merck Index | 15, 8657 |
Solubility Information | Solubility in water: 219 g/100mL (20°C). Other solubilities: soluble in alcohol, slightly soluble in acetone and ether |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement May intensify fire; oxidizer. May be corrosive to metals. Causes severe skin burns and eye damage. Very toxic to aquatic life with long lasting effects. |
Precautionary Statement | GHS P Statement Keep/Store away from clothing/ combustible materials. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off im |
EINECSNumber | 231-853-9 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction