Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver nitrate, ACS, 99.9+% (metals basis)

Catalog No. AA1141436
Click to view available options
Quantity:
5 g
25 g
100 g
500 g

Silver nitrate, AgNO3, CAS Number-7761-88-8, silvernitrate, silver i nitrate, argenti nitras, nitric acid silver i salt, nitrate d'argent, lunar caustic, argerol, silver mononitrate, silver nitrate, silbernitrat, 100g, >99.9% (metals basis), CHEBI:32130, 4.352 g/mL | CAS: 7761-88-8 | AgNO3 | 169.87 g/mol

Silver nitrate is employed in Silver plating, photography, manufacturing of other silver compounds, mirrors, coloring porcelain, and staining in histology. It finds use as a stain in scanning electron microscopy, and a key ingredient in several silver-based antiseptic and antibacterial pharmaceutical compositions. Silver(I) nitrate is used to separate mixtures of alkenes by selective absorption, and for deprotection and oxidation reactions in organic synthesis. It is widely used as an analytical reagent owing to its immediate reaction with halide ions forming insoluble precipitate of silver(I) halides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Melting Point 212°C
Physical Form Crystalline
Quantity 500 g
Assay Percent Range >99.9% (metals basis)
Solubility Information Freely soluble in water,alcohol and aqueous ammonia. Slightly soluble in ether.
Formula Weight 169.87
Grade ACS Reagent
Sensitivity Light sensitive
Density 4.352 g/mL
Chemical Name or Material Silver nitrate, For AAS analysis
Hazard Category H272-H290-H314-H335-H373
Precautionary Statement P220-P221-P234-P260-P262-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 5.1; Packaging Group: II
EINECSNumber 231-853-9
RTECSNumber VW4725000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.