Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Nitrate-Nitric Acid Reagent, for Sulfate Determination, Ricca Chemical
SDP

Catalog No. 70454 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
7045-4 120 mL
7045-16 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 7045-4 Supplier Ricca Chemical Company Supplier No. 70454
Only null left
Add to Cart
Add to Cart

APHA/EPA for Sulfate Testing

Specifications

pH Range <1
Volume (Metric) 120 mL
Packaging Amber Glass Bottle
Quantity 120 mL
Test Method APHA/EPA for Sulfate
Type Silver Nitrate-Nitric Acid Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.