Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Silver Nitrate-Nitric Acid Reagent, for Sulfate Determination, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
120 mL
500 mL
Specifications
Specifications
| pH Range | <1 |
| Volume (Metric) | 500 mL |
| Packaging | Amber Glass Bottle |
| Quantity | 500 mL |
| Test Method | APHA/EPA for Sulfate |
| Type | Silver Nitrate-Nitric Acid Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction