Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Silver Nitrate Solution in Methanol, Reagents
Click to view available options
:
Quantity: 1 L
Specifications
Specifications
Chemical Name or Material | Silver Nitrate Solution in Methanol |
Boiling Point | 67°C |
Color | Colorless |
CAS | 67-56-1, 7761-88-8, 7732-18-5 |
Physical Form | Liquid |
Quantity | 1 L |
Synonym | Nitric Acid silver salt |
Grade | Reagent |
Packaging | Glass Bottle |
Specific Gravity | 0.8 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction