Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver perchlorate monohydrate, 99.9% (metals basis)

Catalog No. p-7048836
Click to view available options
Quantity:
2 g
10 g
50 g

CAS: 14242-05-8 | AgClO4 | 207.31 g/mol

Silver perchlorate can be used in the manufacturing of explosives. It is generally used as a source of silver ions during silver nanoparticles synthesis. It is used as a catalyst in organic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14242-05-8
Molecular Formula AgClO4
Molecular Weight (g/mol) 207.31
MDL Number MFCD00149128
InChI Key YDHABVNRCBNRNZ-UHFFFAOYSA-M
Synonym silver perchlorate hydrate, silver perchlorate monohydrate, silver perchlorite, silver perchloric acid, acmc-1brvv, ag.clo4.h2o, silver i perchlorate monohydrate, silver 1+ ion hydrate perchlorate, silver 1+ hydrate perchlorate
PubChem CID 15959344
IUPAC Name silver;perchlorate;hydrate
SMILES [Ag+].[O-][Cl](=O)(=O)=O

Specifications

Melting Point 485°C (decomposition)
Color White
Physical Form Crystalline
Quantity 2 g
UN Number UN1481
Merck Index 14,8523
Solubility Information Soluble in water and organic solvents.
Formula Weight 225.34 (207.32 Anhydrous)
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic; Light sensitive
Density 2.800 g/mL
Assay (metals basis)
Chemical Name or Material Silver perchlorate monohydrate
Show More Show Less
Hazard Category H272-H314
Hazard Statement GHS H Statement
H272-H314-H318
May intensify fire
oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P220-P221-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.
EINECSNumber 232-035-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.