Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Silver shot, 1-7mm (0.08-0.12in), Premion™, 99.999% (metals basis)

Catalog No. AA4739818
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA47398-18 50 g
AA47398-09 10 g
AA47398-30 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA47398-18 Supplier Thermo Scientific™ Supplier No. 4739818
Add to Cart

CAS: 7440-22-4 | Ag | 107.87 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
IUPAC Name silver
SMILES [Ag]

Specifications

Quantity 50 g
Form Solid
Percent Purity 99.999
ShelfLife 30 Years
EINECSNumber 231-131-3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.