Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Standard, 1mL = 1mg Ag, 1000ppm Ag, Ricca Chemical
SDP

Catalog No. 710016
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Amber Poly

Chemical Identifiers

CAS 7761-88-8, 7697-37-2, 7732-18-5
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Packaging Amber Poly
Concentration or Composition Notes 1000ppm Ag
Equivalence 1 mL = 1 mg Ag
Physical Form Liquid
Color Colorless
Quantity 500 mL
Chemical Name or Material Silver Standard
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.