Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Sulfadiazine, Micronized, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18606241 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
10 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail

Catalog No. 18-606-241

Supplier: Spectrum Chemical Manufacturing Corporation S109325GM

S1093, 22199-08-2, C10H9AgN4O2S

Spectrum™ Chemical Silver Sulfadiazine, Micronized, USP is a topical sulfonamide antibacterial used as a topical cream to treat burns. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 22199-08-2
Molecular Formula C10H9AgN4O2S
Molecular Weight (g/mol) 357.14
InChI Key UEJSSZHHYBHCEL-UHFFFAOYSA-N
IUPAC Name silver(1+) 4-{[(pyrimidin-2-yl)azanidyl]sulfonyl}aniline
SMILES [Ag+].NC1=CC=C(C=C1)S(=O)(=O)[N-]C1=NC=CC=N1

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 98 to 102%
Grade USP
Loss on Drying 0.005
Packaging Amber Glass Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.