Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Sulfate, ACS Reagent, Solstice

Catalog No. 6001352 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
10 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-013-52 10 g
60-013-53 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-013-52 Supplier Solstice Supplier No. 22567310G

ACS Reagent, 99%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-26-5
Molecular Formula Ag2H2O4S
Molecular Weight (g/mol) 313.81
MDL Number MFCD00003407
InChI Key YPNVIBVEFVRZPJ-UHFFFAOYSA-N
Synonym silver sulfate, disilver sulfate, disilver 1+ sulfate, unii-8qg6hv4zpo, disilver 1+ sulphate, sulfuric acid, disilver 1+ salt, 8qg6hv4zpo, disilver 1+ ion sulfate, sulfuric acid, silver 1+ salt 1:2, sulfuric acid disilver i salt
PubChem CID 159865
IUPAC Name disilver(1+) sulfuric acid
SMILES [Ag+].[Ag+].OS(O)(=O)=O

Specifications

Quantity 10 g
Linear Formula Ag2SO4
UN Number 3077
Formula Weight 311.8g/mol
Percent Purity 99%
Grade ACS Reagent
Chemical Name or Material Silver Sulfate
Hazard Statement P280-P305 + P351 + P338 + P310
Precautionary Statement H318-H410
EINECSNumber 233-653-7
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.