Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Silver sulfide, 99.9+% (metals basis)
Click to view available options
Quantity:
5 g
25 g
Description
Silver sulfide is useful as a photosensitizer in photography, Intermediates, chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 825°C |
| Color | Gray to Black |
| Physical Form | Crystalline Powder |
| Quantity | 25 g |
| UN Number | UN3077 |
| Merck Index | 14,8530 |
| Solubility Information | Soluble in aq. HCN,aq. citric acid with KNO3. Insoluble in acids,alkalies,aqueous ammoniums. |
| Formula Weight | 247.8 |
| Percent Purity | ≥99.9% |
| Odor | Odorless |
| Show More |
Safety and Handling
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
| EINECSNumber | 244-438-2 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction