Alfa Aesar™ Silver sulfide, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 4545814

Quantity 25g

 View more versions of this product

Catalog No. AA4545814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 21548-73-2
    Molecular Formula Ag2S
    Formula Weight 247.8
    MDL Number MFCD00003406
    UN Number UN3077
    Merck Index 14,8530
    Chemical Name or Material Silver sulfide

    Specifications

    Formula Weight 247.8
    Particle Size 6mm & down
    Quantity 25g
    TSCA TSCA
    Odor Odorless
    CAS 21548-73-2
    Merck Index 14,8530
    Color Dark gray to black
    Density 7.317g/mL
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Physical Form Pieces
    Molecular Formula Ag2S
    MDL Number MFCD00003406
    UN Number UN3077
    Melting Point 825°C
    Chemical Name or Material Silver sulfide
    Assay 99.999%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit