Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver tetrafluoroborate, 99%

Catalog No. AC197090100
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Plastic bottle

Catalog No. AC197090100

Supplier: Thermo Scientific Chemicals 197090100

This product is not available for sale online. Please call Customer Service at 1-800-766-7000 or send an email to cs.webemail@thermofisher.com for a quote.

CAS: 14104-20-2 | AgBF4 | 194.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14104-20-2
Molecular Formula AgBF4
Molecular Weight (g/mol) 194.67
MDL Number MFCD00003408
InChI Key CCAVYRRHZLAMDJ-UHFFFAOYSA-N
Synonym silver tetrafluoroborate, silver borofluoride, silver fluoborate, silver fluoroborate, silver i tetrafluoroborate, borate 1-, tetrafluoro-, silver 1+, silver tetrafluroborate, acmc-209clm
PubChem CID 159722
SMILES [Ag+].F[B-](F)(F)F

Specifications

Color Beige
Physical Form Crystalline Powder
Quantity 10 g
Assay Percent Range 43.92% min. (BF4)
Linear Formula AgBF4
Solubility Information Solubility in water: soluble.
IUPAC Name silver(1+) tetrafluoroboranuide
Formula Weight 194.67
Percent Purity 99%
Packaging Plastic bottle
Chemical Name or Material Silver tetrafluoroborate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 237-956-5
RTECSNumber ED2875000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.