Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Silver tetrafluoroborate, 99%
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Beige |
Physical Form | Crystalline Powder |
Quantity | 50 g |
Assay Percent Range | 43.92% min. (BF4) |
Linear Formula | AgBF4 |
Solubility Information | Solubility in water: soluble. |
IUPAC Name | silver(1+) tetrafluoroboranuide |
Formula Weight | 194.67 |
Percent Purity | 99% |
Packaging | Plastic bottle |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. |
Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with wat |
EINECSNumber | 237-956-5 |
RTECSNumber | ED2875000 |
TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction