Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver thiocyanate, Thermo Scientific Chemicals

Catalog No. p-4824942
Click to view available options
Quantity:
10 g

For Preparation of Nonmetallic Thiocyanates, Analytical Reagent, in pHotographic Emulsions

Silver thiocyanate is applied in the preparation of nonmetallic thiocyanates, analytical reagent, in photographic emulsions and as an organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1701-93-5
Molecular Formula CHAgNS
Molecular Weight (g/mol) 166.954
MDL Number MFCD00003416
InChI Key KFDDCZCFSUJOBJ-UHFFFAOYSA-N
Synonym silver 1+ ion thiocyanate ion
PubChem CID 16211904
IUPAC Name silver;thiocyanic acid
SMILES C(#N)S.[Ag]

Specifications

Melting Point (decomposition)
Color White
Physical Form Crystalline
Quantity 10 g
Solubility Information Very slightly soluble in water. Insoluble in ethanol,acetone,acid. Soluble in NH4OH
Formula Weight 165.95
Odor Odorless
Sensitivity Light sensitive
Appearance White crystals
Chemical Name or Material Silver thiocyanate
Hazard Category H302+H312+H332
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation DOT Class: 9, Packaging Group: III
EINECSNumber 216-934-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.