Silver thiocyanate, Alfa Aesar™

For Preparation of Nonmetallic Thiocyanates, Analytical Reagent, in pHotographic Emulsions

Manufacturer: Alfa Aesar 1189709

Quantity 10g

 View more versions of this product

Catalog No. AA1189709

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1701-93-5
    Physical Form Crystalline
    Molecular Formula CAgNS
    Formula Weight 165.95
    MDL Number MFCD00003416
    Chemical Name or Material Silver thiocyanate

    Specifications

    Formula Weight 165.95
    Quantity 10g
    Color White
    DOT Information DOT Class: 9, Packaging Group: III
    EINECS Number 216-934-9
    Physical Form Crystalline
    Odor Odorless
    Molecular Formula CAgNS
    Appearance White crystals
    MDL Number MFCD00003416
    CAS 1701-93-5
    Health Hazard 1 H400-H410-H302-H312-H332: Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled.
    Chemical Name or Material Silver thiocyanate

    H400-H410-H302-H312-H332: Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit