Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver tungsten oxide, 99% (metals basis)

Catalog No. aa3966109
Click to view available options
Quantity:
10 g
50 g

CAS: 13465-93-5 | Ag2O4W | 463.57 g/mol

Silver tungsten oxide is used in electronic and chemical industries. It is also used as a biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13465-93-5
Molecular Formula Ag2O4W
Molecular Weight (g/mol) 463.57
MDL Number MFCD00014147
InChI Key JFUVYDSGHRJJDO-UHFFFAOYSA-N
Synonym Silver tungstate

Specifications

Color Yellow
Quantity 10 g
Odor Odorless
Sensitivity Light sensitive
Form Powder
Linear Formula Ag2WO4
Solubility Information Insoluble in water; Soluble in potassium cyanide, ammonium hydroxide and nitric acid
Formula Weight 463.58
Percent Purity 99%
Chemical Name or Material Silver tungsten oxide
EINECSNumber 236-708-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.