Learn More
Description
- Unique five-layer construction provides chemical protection
- Ideal for hospitals, agriculture, military and hazardous material handling
- Easily worn under heavy-duty outer gloves
- Use as a primary glove where there is minimal risk of tearing or puncturing
- Resistant to over 280 different chemicals, including alcohols, aliphatics, aromatics, chlorines, ketones and esters
- Does not contain latex or chemical accelerators
Specifications
Specifications
| Certifications/Compliance | OSHA 29 CFR 1915.157 |
| Certifications/Compliance (Europe) | EN 420, EN 374-3 |
| Color | Silver |
| Hand | Ambidextrous |
| Latex Free | Yes |
| Material | Polyethylene, EVA |
| Size | 8 |
| Thickness (English) | 2.7 mil |
| Thickness (Metric) | 0.06 mm |
| EN388 Standard | EN 388 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.