Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ SilverShield™ SSG Gloves

Catalog No. 18999431E Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Length (English):
14.5 in.
29 in.
Length (Metric):
37 cm
74 cm
Size:
6
7
8
9
10
11
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Length (Metric) Size
18-999-431E 14.5 in. 37 cm 10
18-999-431A 14.5 in. 37 cm 6
18-999-431B 14.5 in. 37 cm 7
18-999-431C 14.5 in. 37 cm 8
18-999-431D 14.5 in. 37 cm 9
18-999-431F 14.5 in. 37 cm 11
19-150-0892 29 in. 74 cm 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 18-999-431E Supplier Honeywell™ Supplier No. SSG/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Unique five-layer fabric is resistant to over 280 different chemicals including alcohols, aliphatics, aromatice, chlorines, ketones and esters.

  • Unique five-layer construction provides chemical protection
  • Ideal for hospitals, agriculture, military and hazardous material handling
  • Easily worn under heavy-duty outer gloves
  • Use as a primary glove where there is minimal risk of tearing or puncturing
  • Resistant to over 280 different chemicals, including alcohols, aliphatics, aromatics, chlorines, ketones and esters
  • Does not contain latex or chemical accelerators

Specifications

Certifications/Compliance OSHA 29 CFR 1915.157
Certifications/Compliance (Europe) EN 420, EN 374-3
Color Silver
Hand Ambidextrous
Latex Free Yes
Material Polyethylene, EVA
Size 10
Thickness (English) 2.7 mil
Thickness (Metric) 0.06 mm
EN388 Standard EN 388
EN ISO 374 - 5 Bacteria, Fungi only
Glove Cuff Style Straight
Grip Design Smooth
Lining Unlined
Model SSG/10
Series SilverShield™ SSG
Length (English) 14.5 in.
Length (Metric) 37 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.