Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113657
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113657 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113657 Supplier Invitrogen™ Supplier No. PA5113657
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MKEKSKNAAR TRREKENSEF YELAKLLPLP SAITSQLDKA SIIRLTTSYL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

SIM1 and SIM2 genes are Drosophila single-minded (sim) gene homologs. SIM1 transcript was detected only in fetal kidney out of various adult and fetal tissues tested. Since the sim gene plays an important role in Drosophila development and has peak levels of expression during the period of neurogenesis, it was proposed that the human SIM gene is a candidate for involvement in certain dysmorphic features (particularly the facial and skull characteristics), abnormalities of brain development, and/or mental retardation of Down syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIM1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SIM1
Gene Accession No. P81133
Gene Alias BHLHE14; bHLH-PAS transcription factor; Class E basic helix-loop-helix protein 14; mSIM1; SIM bHLH transcription factor 1; SIM bHLH transcription factor 1a; sim1; sim1a; single-minded 1; single-minded family bHLH transcription factor 1; single-minded family bHLH transcription factor 1a; single-minded homolog 1; single-minded homolog 1 (Drosophila); single-minded homolog 1-A; single-minded-like 1
Gene Symbols SIM1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human SIM1 (aa 1-50).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6492
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.