Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIM1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5116187

Catalog No. PIPA5116187


Only null left
Add to Cart

Description

Description

SIM1 Polyclonal Antibody for Western Blot, IHC (P)

SIM1 and SIM2 genes are Drosophila single-minded (sim) gene homologs. SIM1 transcript was detected only in fetal kidney out of various adult and fetal tissues tested. Since the sim gene plays an important role in Drosophila development and has peak levels of expression during the period of neurogenesis, it was proposed that the human SIM gene is a candidate for involvement in certain dysmorphic features (particularly the facial and skull characteristics), abnormalities of brain development, and/or mental retardation of Down syndrome.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SIM1
Polyclonal
Unconjugated
SIM1
BHLHE14; bHLH-PAS transcription factor; Class E basic helix-loop-helix protein 14; mSIM1; SIM bHLH transcription factor 1; SIM bHLH transcription factor 1a; sim1; sim1a; single-minded 1; single-minded family bHLH transcription factor 1; single-minded family bHLH transcription factor 1a; single-minded homolog 1; single-minded homolog 1 (Drosophila); single-minded homolog 1-A; single-minded-like 1
Rabbit
Antigen affinity chromatography, Protein A
RUO
6492
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.43 mg/mL
PBS with 0.09% sodium azide; pH 7.4
P81133
SIM1
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human SIM1.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.