Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537887
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537887 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537887 Supplier Invitrogen™ Supplier No. PA537887
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with mouse, rat, dog, pig and cow based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 2,000.

SIM1 and SIM2 genes are Drosophila single-minded (sim) gene homologs. SIM1 transcript was detected only in fetal kidney out of various adult and fetal tissues tested. Since the sim gene plays an important role in Drosophila development and has peak levels of expression during the period of neurogenesis, it was proposed that the human SIM gene is a candidate for involvement in certain dysmorphic features (particularly the facial and skull characteristics), abnormalities of brain development, and/or mental retardation of Down syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIM1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene SIM1
Gene Accession No. P81133
Gene Alias BHLHE14; bHLH-PAS transcription factor; Class E basic helix-loop-helix protein 14; mSIM1; SIM bHLH transcription factor 1; SIM bHLH transcription factor 1a; sim1; sim1a; single-minded 1; single-minded family bHLH transcription factor 1; single-minded family bHLH transcription factor 1a; single-minded homolog 1; single-minded homolog 1 (Drosophila); single-minded homolog 1-A; single-minded-like 1
Gene Symbols SIM1
Host Species Goat
Immunogen Peptide with sequence C-SGDRYRTEQYQS, from the internal region of SIM1 (aa 545-557).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6492
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.