Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIM2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981814
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1814 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1814 Supplier Bioss Supplier No. BS11951R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene represents a homolog of the Drosophila single-minded (sim) gene, which encodes a transcription factor that is a master regulator of neurogenesis. The encoded protein is ubiquitinated by RING-IBR-RING-type E3 ubiquitin ligases, including the parkin RBR E3 ubiquitin protein ligase. This gene maps within the so-called Down syndrome chromosomal region, and is thus thought to contribute to some specific Down syndrome phenotypes. Alternative splicing of this gene results in multiple transcript variants.

Specifications

Antigen SIM2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SIM2
Gene Accession No. Q14190
Gene Alias BHLHE15; Class E basic helix-loop-helix protein 15; HMC13F06; HMC29C01; mSIM; SIM; SIM bHLH transcription factor 2; SIM transcription factor; SIM2; single-minded 2; single-minded family bHLH transcription factor 2; Single-minded homolog 2; single-minded homolog 2 (Drosophila); single-minded protein; transcription factor SIM2
Gene Symbols SIM2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SIM2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6493
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.