Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIMC1 Polyclonal Antibody
SIMC1 Polyclonal Antibody
Supplier: Thermo Scientific PA558153
Description
SIMC1 Polyclonal Antibody for ICC/IF

Specifications
SIMC1 | |
Polyclonal | |
Unconjugated | |
SIMC1 | |
4732471D19Rik; C2H5orf25; C4H5orf25; C5orf25; FLJ44216; oocyte maturation associated 1; OOMA1; platform element for inhibition of autolytic degradation; PLEIAD; Simc1; SUMO interacting motifs containing 1; SUMO-interacting motif-containing protein 1; SUMO-interacting motifs containing 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
375484 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NDZ2 | |
SIMC1 | |
Recombinant protein corresponding to Human SIMC1. Recombinant protein control fragment (Product #RP-96929). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction