Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIMC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565196
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565196 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565196 Supplier Invitrogen™ Supplier No. PA565196
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YVIDLTRAEGE NRPIATLDLT LEPVTPSQRE PTSLQTCASL SGKAVMEGQV DRSSQPTARR LINSDPVDLD LVEE Highest antigen sequence identity to the following orthologs - mouse 77%, rat 72%.

SIMC1 (SUMO Interacting Motifs Containing 1) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includeSUMO polymer binding. An important paralog of this gene isSKIDA1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIMC1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SIMC1
Gene Accession No. Q8NDZ2
Gene Alias 4732471D19Rik; C2H5orf25; C4H5orf25; C5orf25; FLJ44216; oocyte maturation associated 1; OOMA1; platform element for inhibition of autolytic degradation; PLEIAD; Simc1; SUMO interacting motifs containing 1; SUMO-interacting motif-containing protein 1; SUMO-interacting motifs containing 1
Gene Symbols SIMC1
Host Species Rabbit
Immunogen Recombinant Human SIMC1. Recombinant protein control fragment (Product #RP-105939).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 375484
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.