Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIMC1 Polyclonal Antibody
SIMC1 Polyclonal Antibody
Supplier: Thermo Scientific PA570170
Description
SIMC1 Polyclonal Antibody for Western Blot

Specifications
SIMC1 | |
Polyclonal | |
Unconjugated | |
SIMC1 | |
4732471D19Rik; C2H5orf25; C4H5orf25; C5orf25; FLJ44216; oocyte maturation associated 1; OOMA1; platform element for inhibition of autolytic degradation; PLEIAD; Simc1; SUMO interacting motifs containing 1; SUMO-interacting motif-containing protein 1; SUMO-interacting motifs containing 1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
319719 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
E9Q6E9 | |
SIMC1 | |
synthetic peptide directed towards the middle region of human SIMC1 (aa 1236-1285). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction