Learn More
SIMC1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305759AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
SIMC1 (SUMO Interacting Motifs Containing 1) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includeSUMO polymer binding. An important paralog of this gene isSKIDA1.Specifications
SIMC1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q8NDZ2 | |
SIMC1 | |
between 25 and 75 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunoprecipitation | |
Unconjugated | |
SIMC1 | |
C5orf25, chromosome 5 open reading frame 25, FLJ44216, oocyte maturation associated 1, OOMA1, platform element for inhibition of autolytic degradation, PLEIAD, SUMO-interacting motifs containing 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
375484 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.