Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Learning Resources Simple Tape Measure

Click to view available options
Class Size:
1 Student
Description
- Introduce your youngest students to measuring with this real, manual-wind tape measure.
- This tool can measure up to 4 feet and features inches and feet on one side, and centimeters and meter on the other.
Specifications
Specifications
| Grade Level | Elementary, Intermediate - Middle School, High School |
| Grade | K to 12 |
| Class Size | 1 Student |
| Product Type | Measurement Tape |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction