Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simport™ Scientific Amplitube™ PCR Reaction Strips

Catalog No. 22038190 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Blue
Green
Natural
Red
Yellow
Closure Type:
Domed Cap
Flat Cap
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Closure Type
22-038-190 Natural Domed Cap
22-039-101 Natural Flat Cap
23-045-236 Blue Flat Cap
23-045-237 Green Flat Cap
23-045-238 Red Flat Cap
23-045-239 Yellow Flat Cap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 missing translation for 'options'
This item is not returnable. View return policy
Catalog No. 22-038-190 missing translation for 'mfr' Simport™ Scientific missing translation for 'supplierNo' T3203N
Add to Cart
Add to Cart
This item is not returnable. View return policy

Made of polypropylene. Amplitube™ PCR Reaction Strips are manufactured under the most stringent conditions to attain the highest quality standards.

  • Available with either flat (needle pierceable) or dome-topped individually attached hinged caps
  • Hinged caps have integral seal that prevents contamination of lid surface
  • Ultra-thin sidewalls and bottoms for more uniform and efficient temperature transfers
  • Positive sealing fully protects contents from evaporation during thermal cycle
  • Strip fits standard well spacing and can also be used with 8-channel hand-held pipettors
  • Easily opened and closed with one hand
  • Autoclavable
  • Choice of colorless and four non-cytotoxic and non-metallic colors
  • Packed in tamper-proof resealable bags

Specifications

Color Natural
Volume (Metric) 0.2 mL
Quantity 125 Pk.
For Use With (Equipment) Standard well spacing and can also be used with 8-channel hand-held pipettors
Sterility Non-sterile
Material Polypropylene
Closure Type Domed Cap
Autoclavable Autoclavable
Format Tube
Product Type 8-Strip PCR Reaction Tube
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.