Description
- Made of acetal polymer and resistant to chemical action of histological solvents
- Fit securely in microtome chuck adapters
- Accepts standard metal lids that keep specimens safe during processing
- Slanted writing surface for easy sample identification
- Available in three colors
Certifications
CE marked
Specifications
Specifications
| Color | Peach |
| Material | Acetal Polymer |
| Product Type | Embedding Cassette |