Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simport™ Scientific Screw Caps for T501 Sample Tubes

Catalog No. 0567993 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Dark Green
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
05-679-93 Dark Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-679-93 Supplier Simport™ Scientific Supplier No. T501DG
Add to Cart
Add to Cart

Screw Caps for T501 Sample Tubes are available either with or without a silicone washer between the cap and the tube to ensure a positive leakproof seal at all temperatures. The caps feature a long skirt and a super-fast thread design that allows it to be removed or sealed with a single turn.

  • Made of polypropylene
  • Available with or without silicone washer
  • Long skirt and super fast thread design for easy seal
  • Autoclavable

Specifications

Includes Lip seal
Product Type Screw Cap
Autoclavable Autoclavable
Material Polypropylene
Color Dark Green
Sterility Non-sterile
For Use With (Equipment) Sample Tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.