Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simport™ Scientific Nonsterile Screw Caps

Catalog No. 0567996 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Blue
Natural
Yellow
Includes:
Lip seal
Silicone washer
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Includes
05-679-96 Yellow Lip seal
22-038-265 Natural Silicone washer
22-039-102 Blue Silicone washer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 05-679-96 Supplier Simport™ Scientific Supplier No. T501Y
Add to Cart
Add to Cart

Nonsterile Screw Caps are available either with or without a silicone washer between the cap and the tube to ensure a positive leakproof seal at all temperatures. The cap features a long skirt and a super-fast thread design that allows it to be removed or sealed with a single turn.

  • Made of polypropylene
  • Available with or without silicone washer
  • Long skirt and super fast thread design for easy seal
  • Autoclavable

Specifications

Includes Lip seal
Product Type Screw Cap
Autoclavable Autoclavable
Material Polypropylene
Color Yellow
Sterility Non-sterile
For Use With (Equipment) Sample Tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.