Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simport™ Scientific Capinsert™ For Cryovial Tubes

Catalog No. 23045225 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Assorted
Blue
Burnt Orange
Green
Lilac
Pink
Red
Tan
White
Yellow
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-045-225 White
22-038-503 Burnt Orange
22-038-501 Yellow
22-038-504 Lilac
22-038-502 Pink
23-045-226 Blue
23-045-227 Red
23-045-228 Green
23-045-229 Assorted
23-045-230 Tan
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 23-045-225 Supplier Simport™ Scientific Supplier No. T3121
Add to Cart
Add to Cart

The Capinsert™ For Cryovial Tubes inserts fit precisely into the cap of the Cryovial™ for color identification.

  • Color coded inserts
  • Made of polypropylene
  • For Cryovial tubes

Specifications

Color White
Material Polypropylene
For Use With (Equipment) Cryovial™ Tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.