Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simport™ Scientific T100 BioTube™ Racks

Catalog No. 055006 Shop All Simport Scientific Products
Change view
Click to view available options
Product Type:
12-strip Tube Rack
96-plain Tube Rack
Color:
Blue
Red
White
Sterility:
Non-sterile
Sterile
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Color Sterility
05-500-6 96-plain Tube Rack Red Sterile
05-500-8 12-strip Tube Rack Blue Non-sterile
05-500-31 12-strip Tube Rack White Sterile
05-500-2 96-plain Tube Rack Red Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 05-500-6 Supplier Simport™ Scientific Supplier No. T1002R
Add to Cart
Add to Cart

The T100 BioTube™ Racks rack with the standard 96-well on-center spacing of tubes offers a color-coding system using colored interchangeable plastic grids.

  • Made of polypropylene
  • Free of RNase, DNase, Pyrogen, and DNA
  • Convenient carrying handles on both sides
  • Cover, grid plate, and base are keyed to prevent misalignment
  • Easy to read ID numbers and letters
  • Grid plate stands on 4 legs and can be placed on a lab counter or in a water bath
  • These racks and tubes are also ideal for storing, freezing, and transporting reagents and specimens

Specifications

Closure Material Polypropylene
Color Red
Material Polypropylene
For Use With HTLV-III testing, bacterial and hybridoma cell uptake studies, cell harvesting
Includes Rack with 96 plain individual tubes
Quantity 10/Cs.
Sterility Sterile
Autoclavable Autoclavable
Product Type 96-plain Tube Rack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.