Learn More
Simport™ Scientific T100 Caps
Catalog No. 0550027 Shop All Simport Scientific Products
Change view
Click to view available options
Autoclavable:
Not Autoclavable
Catalog No. Autoclavable
05-500-27 Not Autoclavable
1 options
Catalog No. 05-500-27 Supplier Simport™ Scientific Supplier No. T10040
Add to Cart
Add to Cart
T100 Caps are made of polyethylene and are not autoclavable. Tubes with a cap in place will hold 1.1 mL.
  • Caps made of polyethylene - not autoclavable
  • Free of RNase, DNase, Pyrogen, and DNA
  • Available as individual, 8-strip, and 12-strip configuration
  • Volume with cap is 1.1 mL
  • Tubes sold separately
Specifications
Material Polyethylene
For Use With HTLV-III testing, bacterial and hybridoma cell uptake studies, cell harvesting
Sterility Non-sterile
Autoclavable Not Autoclavable
Product Type 12-plug Cap Strip