Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simport™ Scientific CytoSep™ Filter Papers for Shandon Cytospin 4™ Centrifuges

Catalog No. 22045305 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Tan
White
For Use With (Equipment):
Double Funnel
Single Funnel
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color For Use With (Equipment)
22-045-305 White Single Funnel
22-045-306 Tan Single Funnel
22-045-309 White Double Funnel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-045-305 Supplier Simport™ Scientific Supplier No. M965FW
Add to Cart
Add to Cart

CytoSep™ Filter Papers for Shandon Cytospin 4™ Centrifuges for double and single funnels fitting Thermo Scientific™ Shandon™ Cytospin 4™ Cytocentrifuges.

  • Available in white and tan

Specifications

Color White
For Use With (Equipment) Single Funnel
Type Filter Paper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.