Learn More
Simport™ Scientific CytoSep™ Fluid Chamber for Sakura Cyto-Tek Cytocentrifuge
Catalog No. 22045317 Shop All Simport Scientific Products
Change view
Click to view available options
Volume (Metric):
1 mL
Catalog No. Volume (Metric)
22-045-317 1 mL
1 options
Catalog No. 22-045-317 Supplier Simport™ Scientific Supplier No. M9631
Add to Cart
Add to Cart
Disposable CytoSep™ Fluid Chamber for Sakura Cyto-Tek Cytocentrifuge offers lower risk of contamination than reusable funnels.
  • Offers 1 mL fluid chamber component
Specifications
Disposable Single-use
For Use With (Equipment) Sakura Cyto-Tek
Includes All Components
Type Fluid Chamber Only
Volume (Metric) 1 mL