Learn More
Description
- Available in white and tan
Specifications
Specifications
| Color | White |
| For Use With (Equipment) | Wescor Cyto-System |
| Includes | White Filter Paper and Cap |
| Type | Single Sample Chamber |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.