Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simport™ Scientific Sample Tubes

Catalog No. p-4272599 Shop All Simport Scientific Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
22-038-410 Non-sterile
22-038-409 Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-038-410 Supplier Simport™ Scientific Supplier No. C5722
Only null left
Add to Cart
Add to Cart

Sample Tubes for transport and storage of urine, sputum and most liquids or particulate samples.

  • Tube made of polypropylene
  • Cap made of polyethylene
  • Conical bottom tubes with self-standing base graduated
  • Chemically resistant and shatterproof
  • Supplied with a leakproof screw cap
  • Molded-in graduations make them easy to read
  • For transporting liquid sample/hazardous material

Specifications

Diameter (Metric) 25.3 mm
Product Type Sample Tube
Closure Color White
Autoclavable Autoclavable
Capacity (Metric) 30 mL
Material Polypropylene
For Use With (Application) Transport and storage of urine, sputum and most liquids or particulate samples
Length (Metric) 111 mm
No. per Case 500/Cs.
No. per Pack 100/Bag
Sterility Non-sterile
Closure Material Polyethylene
Capacity (English) 1.01 oz.
Closure Type Screw Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.