Learn More
Simport™ Scientific Septum Screw Cap for Sample Tubes
Catalog No. 22040400 Shop All Simport Scientific Products
Change view
Click to view available options
Autoclavable:
Autoclavable
Catalog No. Autoclavable
22-040-400 Autoclavable
1 options
Catalog No. 22-040-400 Supplier Simport™ Scientific Supplier No. T504AQX
Add to Cart
Add to Cart
The Septum Screw Cap for Sample Tubes is made to fit and be used with Simport sample tubes (T501 series).
  • Made of polypropylene
  • Septum made of chemically resistant PTFE outside and silicone inside
  • Also acts as O-ring
  • Pierceable with pipet tips and syringe needles
Specifications
Product Type Septum Screw Cap
Autoclavable Autoclavable
Material Polypropylene
For Use With (Equipment) Simport Sample Tubes