Learn More
Simport™ Scientific SLIMSETTE™ M509SL Tissue Processing/Embedding Cassettes in QuickLoad™ Sleeves
Shop All Simport Scientific ProductsDescription
- Made of acetal polymer
- Transparent sleeve allows clear view of cassettes during printing
- Efficient flow-through slots maximize fluid exchange and ensure proper drainage
- Lid can be opened and closed as often as necessary, and will always relock securely
- Large labeling area for easy identification
- Anterior writing area at a 45° angle
Specifications
Specifications
| Pore Style | Slots |
| Writing Face Angle | 45° |
| Closure Type | Integral Lid |
| Color | Tan |
| Material | Acetal Polymer |
| Product Type | Tissue Cassette |