Learn More
Simport™ Scientific SLIMSETTE™ M510SL Biopsy Processing/Embedding Cassettes in Quickload™ Sleeves
Catalog No. 23045214 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Aqua
Blue
Green
Lilac
Orange
Peach
Pink
Tan
White
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-045-214 Lilac
23-045-216 Aqua
23-045-215 Orange
23-045-206 White
23-045-212 Tan
23-045-207 Pink
23-045-211 Peach
23-045-210 Blue
23-045-208 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 23-045-214 Supplier Simport™ Scientific Supplier No. M51010SL
Add to Cart
Add to Cart
SLIMSETTE™ M510SL Biopsy Processing/Embedding Cassettes in Quickload™ Sleeves are a new generation of cassettes, prepackaged for use in automatic cassette labeling instruments.
  • Specially designed to hold biopsy specimens during the embedding process
  • Made of acetal polymer
  • Transparent sleeve allows clear view of cassettes during printing
  • Efficient flow-through slots maximize fluid exchange and ensure proper drainage
  • Lid can be opened and closed as often as necessary, and will always relock securely
  • Large labeling area for easy identification
  • Anterior writing area at a 45° angle
Specifications
Pore Style Slots
Writing Face Angle 45°
Closure Type Integral Lid
Color Lilac
Material Acetal Polymer
Product Type Biopsy Cassette