Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simport™ Scientific Tamper Evident Self-Standing Transport Tubes

Catalog No. 22040240 Shop All Simport Scientific Products
Change view
Click to view available options
Capacity (Metric):
5 mL
7 mL
10 mL
12 mL
Graduated:
No
Yes
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Graduated
22-040-240 12 mL Yes
22-040-237 5 mL Yes
22-040-239 10 mL Yes
22-040-247 7 mL No
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 22-040-240 Supplier Simport™ Scientific Supplier No. T55212ATTP
Only null left
Add to Cart
Add to Cart

Tamper Evident Self-Standing Transport Tubes for all applications requiring tamper evident seal for utmost security and sample integrity.

  • For secure short and long term storage
  • Provide strength and clarity
  • Non-toxic, non sterile polypropylene made etched or non graduated tubes
  • Exhibit unique design features
  • Unique transport tubes incorporate a seal that needs to be broken in order to open it, which leaves visual indication that the vial has been opened
  • With external threads for providing smooth and uniform inner surface
  • Closure offers unique frangible band design
  • Combination of tamper evident and resistant closure system
  • For clinical trials and expensive reagents in diagnostic kits
  • Caps sold separately

Specifications

Product Type Transport Tube (Self-Standing)
Diameter (Metric) 16.6 mm
Autoclavable Autoclavable
Capacity (Metric) 12 mL
Bottom Shape Conical
Material Polypropylene
For Use With (Application) Storage and transportation of biological materials
Length (Metric) 102 mm
Sterility Non-sterile
Graduated Yes
Packaging Quantity 1000 Pk.
Closure Material Polyethylene
Capacity (English) 0.40 oz.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.