Learn More
Simport™ Scientific Swingsette™ Biopsy Cassettes In Quickload™ Sleeves
Catalog No. 22038650 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-038-650 Green
22-038-648 White
22-038-649 Pink
22-038-653 Peach
22-038-652 Blue
22-038-651 Yellow
22-038-655 Gray
22-038-654 Tan
22-038-656 Lilac
22-038-657 Orange
22-038-658 Aqua
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 22-038-650 Supplier Simport™ Scientific Supplier No. M5184SL
Add to Cart
Add to Cart
Suitable for hoppers accepting plastic sleeves, these Swingsette™ Biopsy Cassettes In Quickload™ Sleeves will load in cassette labeling instruments in one simple operation. Specially designed to hold biopsy specimens during the processing/embedding process as well as in storage cabinets.
  • Made of acetal
  • 1 mm square openings to maximize fluid exchange
  • Sleeved cassettes for use with ThermoFisher printers
  • Anterior writing area at a 45° angle
  • Large tab for convenient and easy opening
  • Covers packaged separately
  • Easy-to-assemble disposable cover with special hinge
Specifications
Color Green
Product Type Biopsy Cassette